Neopentyl glycolbis(4-aminophenyl)ether 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethylpropane 1,3-Bis(4-aminophenyl) Ether, 1,3-Bis(4-aminophenoxy)neopentane, 2,2-Bis[(4-aminophenoxy)methyl]propane

A446067-1G A446067-25G A446067-5G
76725-516EA 17 USD
76725-516 76725-518 76725-520
Neopentyl glycolbis(4-aminophenyl)ether 97%
Neopentyl glycolbis(4-aminophenyl)ether

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₂
MW: 286.38 g/mol
MDL Number: MFCD00671569
CAS Number: 115570-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR