Secnidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol, 1-(2-Methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol

A445585-100MG A445585-10G A445585-25G
76725-510EA 5.31 USD
76725-510 76725-512 76725-514
Secnidazole 98%
Secnidazole

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₃
MW: 185.18 g/mol
MDL Number: MFCD00864656
CAS Number: 3366-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR