Mevastatin 95%

Supplier: Ambeed
Warning

Synonyms: (1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate, Compactin

A445160-10MG A445160-50MG A445160-1G A445160-5G A445160-500G A445160-100G A445160-25G
76725-506EA 5.31 USD
76725-506 76725-508 77572-392 77572-394 77668-994 77668-990 77668-992
Mevastatin 95%
Mevastatin

Precaución: for Research Use Only

Formula: C₂₃H₃₄O₅
MW: 390.52 g/mol
MDL Number: MFCD05662341
CAS Number: 73573-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR