Silver p-toluenesulfonate 97%

Supplier: Ambeed
Warning

A444955-100G A444955-5G
76725-502EA 162 USD
76725-502 76725-504
Silver p-toluenesulfonate 97%
Silver p-toluenesulphonate

Precaución: for Research Use Only

Formula: C₇H₇AgO₃S
MDL Number: MFCD00007511
CAS Number: 16836-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR