Benzyl(methyl)amine 98%

Supplier: Ambeed
Danger

Synonyms: N-Benzylmethylamine, N-Methyl-1-phenylmethanamine, N-Methylbenzylamine

A444580-10G A444580-25G A444580-100G
76725-498EA 5.31 USD
76725-498 76725-500 77601-924
Benzyl(methyl)amine 98%
Benzyl(methyl)amine

Precaución: for Research Use Only

Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
MDL Number: MFCD00008289
CAS Number: 103-67-3
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR