N-Benzyl-N-(4-nitrophenyl)amine 98+%

Supplier: Ambeed
Warning

A443760-1G A443760-250MG
76725-494EA 65 USD
76725-494 76725-496
N-Benzyl-N-(4-nitrophenyl)amine 98+%
N-Benzyl-N-(4-nitrophenyl)amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₂
MDL Number: MFCD00126504
CAS Number: 14309-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR