2,4-Dimethylbenzenesulfonic acid monohydrate 98%

Supplier: Ambeed
Warning

A443737-100G A443737-25G
76725-490EA 11.67 USD
76725-490 76725-492
2,4-Dimethylbenzenesulfonic acid monohydrate 98%
2,4-Dimethylbenzenesulfonic acid monohydrate

Precaución: for Research Use Only

Formula: C₈H₁₂O₄S
MW: 204.24 g/mol
MDL Number: MFCD00673743
CAS Number: 58811-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR