Ruthenium(III) chloride trihydrate 95%

Supplier: Ambeed
Danger

A442445-10G A442445-1G A442445-250MG A442445-25G A442445-5G
76725-472EA 122 USD
76725-472 76725-474 76725-476 76725-478 76725-480
Ruthenium(III) chloride trihydrate 95%
Ruthenium(III) chloride trihydrate

Precaución: for Research Use Only

Formula: Cl₃Ru·3H₂O
MDL Number: MFCD00149845
CAS Number: 13815-94-6
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR