m-Xylylene diisocyanate 98%

Supplier: Ambeed
Danger

A439987-25G A439987-5G
76725-448EA 25 USD
76725-448 76725-450
m-Xylylene diisocyanate 98%
m-Xylylene diisocyanate

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00019917
CAS Number: 3634-83-1
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR