Oxamic acid potassium salt 98%

Supplier: Ambeed
Warning

Synonyms: Potassium Oxamate, Oxamidic Acid Potassium Salt

A437040-25G A437040-5G
76725-440EA 67 USD
76725-440 76725-442
Oxamic acid potassium salt 98%
Oxamic acid potassium salt

Precaución: for Research Use Only

Formula: C₂H₃NO₃
MW: 127.14 g/mol
MDL Number: MFCD00058475
CAS Number: 21141-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR