1-(1-Naphthyl)ethanol 98%

Supplier: Ambeed
Warning

A436354-1G A436354-5G A436354-100G A436354-25G
76725-430EA 32 USD
76725-430 76725-432 77601-884 77668-950
1-(1-Naphthyl)ethanol 98%
1-(1-Naphthyl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MDL Number: MFCD00016806
CAS Number: 1517-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR