N-(2-Diethylaminoethyl)-1-naphthylamine oxalate 95%

Supplier: Ambeed
Warning

A435773-1G A435773-25G A435773-5G
76725-418EA 21 USD
76725-418 76725-420 76725-422
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate 95%
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₄
MDL Number: MFCD00060156
CAS Number: 29473-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR