Potassium-2-formylphenyltrifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(2-formylphenyl)borate, Potassium 2-formylphenyltrifluoroborate

A430931-100MG A430931-1G A430931-250MG A430931-5G
76725-388EA 15 USD
76725-388 76725-390 76725-392 76725-394
Potassium-2-formylphenyltrifluoroborate 98%
Potassium 2-formylphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₅BF₃KO
MW: 212.02 g/mol
MDL Number: MFCD02093979
CAS Number: 192863-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR