N-Methylsuccinimide 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrrolidine-2,5-dione

A430892-10G A430892-1G A430892-25G A430892-5G
76725-376EA 47.75 USD
76725-376 76725-378 76725-380 76725-382
N-Methylsuccinimide 97%
N-Methylsuccinimide

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MW: 113.12 g/mol
MDL Number: MFCD00005517
CAS Number: 1121-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR