Methyl-o-tolylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(o-tolyl)acetate

A430292-100G A430292-1G A430292-25G A430292-5G
76725-364EA 244.01 USD
76725-364 76725-366 76725-368 76725-370
Methyl-o-tolylacetate 95%
Methyl-o-tolylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD00082780
CAS Number: 40851-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR