N-(3-Methoxyphenyl)-2,2-dimethylpropanamide 97%

Supplier: Ambeed
Warning

Synonyms: N-(2,2-Dimethylpropanoyl)-3-(methoxy)aniline

A430225-1G A430225-25G A430225-5G
76725-358EA 23.34 USD
76725-358 76725-360 76725-362
N-(3-Methoxyphenyl)-2,2-dimethylpropanamide 97%
N-(3-Methoxyphenyl)-2,2-dimethylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
MDL Number: MFCD00662896
CAS Number: 56619-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR