Potassium methyltrifluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(methyl)borate, Methyltrifluoroboric acid potassium salt

A429971-100G A429971-10G A429971-1G A429971-250MG A429971-25G A429971-5G
76725-344EA 116 USD
76725-344 76725-346 76725-348 76725-350 76725-352 76725-354
Potassium methyltrifluoroborate 97%
Potassium methyltrifluoroborate

Precaución: for Research Use Only

Formula: CH₃BF₃K
MDL Number: MFCD08276757
CAS Number: 13862-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR