Butyl cyanoacetate 98%

Supplier: Ambeed
Warning

Synonyms: Cyanoacetic acid butyl ester

A428578-100G A428578-25G A428578-500G
76725-318EA 28.65 USD
76725-318 76725-320 76725-322
Butyl cyanoacetate 98%
Butyl cyanoacetate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
MDL Number: MFCD00001941
CAS Number: 5459-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR