Phenyl salicylate 98%

Supplier: Ambeed
Warning

A426987-25G A426987-500G A426987-100G
76725-292EA 13 USD
76725-292 77668-904 77879-929
Phenyl salicylate 98%
Phenyl salicylate

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
MDL Number: MFCD00002213
CAS Number: 118-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR