1,3-Bis(hydroxymethyl)urea 97%

Supplier: Ambeed
Warning

Synonyms: N,N'-Bis(hydroxymethyl)urea, N,N'-Dimethylolurea

A426078-25G A426078-500G
76725-286EA 10.61 USD
76725-286 76725-288
1,3-Bis(hydroxymethyl)urea 97%
1,3-Bis(hydroxymethyl)urea

Precaución: for Research Use Only

Formula: C₃H₈N₂O₃
MW: 120.11 g/mol
MDL Number: MFCD00014414
CAS Number: 140-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR