Potassium tetrathionate 98%

Supplier: Ambeed
Warning

A424907-100G A424907-25G A424907-5G
76725-278EA 152.01 USD
76725-278 76725-280 76725-282
Potassium tetrathionate 98%
Potassium tetrathionate

Precaución: for Research Use Only

Formula: K₂O₆S₄
MDL Number: MFCD00079233
CAS Number: 13932-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR