Pyrrolo[2,1-f][1,2,4]triazin-4-amine 95%

Supplier: Ambeed
Warning

A424610-100G A424610-10G A424610-1G A424610-250MG A424610-25G A424610-5G
76725-266EA 203 USD
76725-266 76725-268 76725-270 76725-272 76725-274 76725-276
Pyrrolo[2,1-f][1,2,4]triazin-4-amine 95%
Pyrrolo[2,1-f][1,2,4]triazin-4-amine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MW: 134.14 g/mol
MDL Number: MFCD08234647
CAS Number: 159326-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR