4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-amine 98%

Supplier: Ambeed
Warning

A420342-100MG A420342-1G A420342-250MG
76725-236EA 12 USD
76725-236 76725-238 76725-240
4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-amine 98%
4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃N
MDL Number: MFCD03424687
CAS Number: 57688-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR