Quinaldine red ≥95% (dye content)

Supplier: Ambeed
Warning

Synonyms: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

A420333-1G A420333-5G
76725-232EA 18.04 USD
76725-232 76725-234
Quinaldine red ≥95% (dye content)
Quinaldine red

Precaución: for Research Use Only

Formula: C₂₁H₂₃IN₂
MW: 430.33 g/mol
MDL Number: MFCD00011968
CAS Number: 117-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR