5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-(Cyclohexylcarbamoyl)-2-fluorophenyl)boronic acid, 5-(Cyclohexylcarbmoyl)-2-fluorobenzeneboronic acid

A418903-1G A418903-250MG
76725-218EA 283.27 USD
76725-218 76725-220
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid 98%
5-(Cyclohexylcarbamoyl)-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇BFNO₃
MW: 265.088 g/mol
MDL Number: MFCD08235096
CAS Number: 874289-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR