Propylamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propan-1-amine hydrochloride, 1-Aminopropane hydrochloride
A418048-100G
A418048-25G
A418048-10G
A418048-5G
76725-212EA
204
USD
76725-212
76725-214
77668-878
77668-880
Propylamine hydrochloride 98%
Propylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₉N MW: 95.57 g/mol |
MDL Number:
MFCD00012911 CAS Number: 556-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...