(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: (-)-2,3:4,6-Di-O-isopropylidene-alpha-L-xylo-2-hexulofuranosonic acid monohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonicacidmonohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate, Dikegulac monohydrate

A412603-100G A412603-25G A412603-5G
76725-182EA 151.71 USD
76725-182 76725-184 76725-186
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₇·H₂O
MW: 274.27 g/mol
MDL Number: MFCD00150517
CAS Number: 68539-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR