N,N-Dimethylnicotinamide 95%

Supplier: Ambeed
Warning

A410473-250MG A410473-1G
76725-176EA 23.34 USD
76725-176 77572-276
N,N-Dimethylnicotinamide 95%
N,N-Dimethylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00059169
CAS Number: 6972-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR