3-Bromobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromophenyl)(phenyl)methanone, m-Bromobenzophenone

A409781-100G A409781-10G A409781-1G A409781-5G
76725-168EA 326.76 USD
76725-168 76725-170 76725-172 76725-174
3-Bromobenzophenone 98%
3-Bromobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉BrO
MW: 261.12 g/mol
MDL Number: MFCD00672015
CAS Number: 1016-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR