4-Nitrobenzyl acetoacetate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrobenzyl 3-oxobutanoate, Acetoacetic Acid 4-Nitrobenzyl Ester

A408526-1000G A408526-100G A408526-10G A408526-500G
76725-158EA 162.32 USD
76725-158 76725-160 76725-162 76725-164
4-Nitrobenzyl acetoacetate 97%
4-Nitrobenzyl acetoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₅
MW: 237.21 g/mol
MDL Number: MFCD00799237
CAS Number: 61312-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR