Pyrophosphoricacidtris(tetrabutylammonium)salt 95%

Supplier: Ambeed
Warning

A408328-1G A408328-250MG A408328-5G
76725-154EA 30 USD
76725-154 76725-156 77551-224
Pyrophosphoricacidtris(tetrabutylammonium)salt 95%
Pyrophosphoricacidtris(tetrabutylammonium)salt

Precaución: for Research Use Only

Formula: C₄₈H₁₀₉N₃O₇P₂
MDL Number: MFCD00210109
CAS Number: 76947-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR