4-Bromo-2-ethylphenol 95%

Supplier: Ambeed
Danger

A408223-100MG A408223-1G A408223-250MG
76725-146EA 47 USD
76725-146 76725-148 76725-150
4-Bromo-2-ethylphenol 95%
4-Bromo-2-ethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD00055489
CAS Number: 18980-21-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR