Potassium (3-methylphenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(m-tolyl)borate, (3-Methylphenyl)trifluoroboric acid potassium salt

A407135-5G A407135-25G
76725-138EA 92 USD
76725-138 77879-892
Potassium (3-methylphenyl)trifluoroborate 98%
Potassium (3-methylphenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃K
MW: 198.04
MDL Number: MFCD04115739
CAS Number: 850623-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR