Potassium hexachlororhodate (III) 98%

Supplier: Ambeed
Warning

Synonyms: Potassium hexachlororhodate(III)

A405898-100MG A405898-1G A405898-250MG
76725-120EA 81.69 USD
76725-120 76725-122 76725-124
Potassium hexachlororhodate (III) 98%
Potassium hexachlororhodate (III)

Precaución: for Research Use Only

Formula: Cl₆K₃Rh
MW: 432.92 g/mol
MDL Number: MFCD00192414
CAS Number: 13845-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR