Sodium hexametaphosphate 97%

Supplier: Ambeed
Warning

Synonyms: Sodium metaphosphate, SHMP

A400494-1000G A400494-100G A400494-25G A400494-500G
76725-088EA 15.92 USD
76725-088 76725-090 76725-092 76725-094
Sodium hexametaphosphate 97%
Sodium hexametaphosphate

Precaución: for Research Use Only

Formula: Na₆P₆O₁₈
MW: 611.77 g/mol
MDL Number: MFCD00136045
CAS Number: 10124-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR