Potassium hexacyanocobaltate(III) 98+%

Supplier: Ambeed
Warning

Synonyms: Tripotassium hexacyanocobaltate(III)

A399980-100G A399980-25G A399980-500G A399980-5G
76725-072EA 70 USD
76725-072 76725-074 76725-076 76725-078
Potassium hexacyanocobaltate(III) 98+%
Potassium hexacyanocobaltate(III)

Precaución: for Research Use Only

Formula: C₆CoK₃N₆
MW: 332.332 g/mol
MDL Number: MFCD00011391
CAS Number: 13963-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR