N-Benzyl-2-(2-nitro-1H-imidazol-1-yl)acetamide 95%

Supplier: Ambeed
Warning

Synonyms: Benznidazole

A398551-1MG A398551-5MG
76725-062EA 16 USD
76725-062 76725-064
N-Benzyl-2-(2-nitro-1H-imidazol-1-yl)acetamide 95%
N-Benzyl-2-(2-nitro-1H-imidazol-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₄O₃
MW: 260.25 g/mol
MDL Number: MFCD00243089
CAS Number: 22994-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR