4-Phenoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A396423-100G A396423-10G A396423-1G A396423-5G
76725-044EA 235 USD
76725-044 76725-046 76725-048 76725-050
4-Phenoxybenzaldehyde 97%
4-Phenoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MDL Number: MFCD00003383
CAS Number: 67-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR