Triethylene glycol monobenzyl ether 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol, 2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethanol
A394990-1G
A394990-25G
A394990-5G
76725-036EA
13.8
USD
76725-036
76725-038
76725-040
Triethylene glycol monobenzyl ether 96%
Triethylene glycol monobenzyl ether
Precaución: for Research Use Only
Formula:
C₁₃H₂₀O₄ MW: 240.30 g/mol |
MDL Number:
MFCD02093556 CAS Number: 55489-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...