Nimorazole 98%

Supplier: Ambeed
Danger

A394939-100MG A394939-10MG A394939-1MG A394939-50MG A394939-1G A394939-250MG A394939-5G
76725-028EA 128.37 USD
76725-028 76725-030 76725-032 76725-034 77601-742 77601-744 77601-746
Nimorazole 98%
Nimorazole

Precaución: for Research Use Only

Formula: C₉H₁₄N₄O₃
MW: 226.23 g/mol
MDL Number: MFCD00866796
CAS Number: 6506-37-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR