Platinum(II) acetylacetonate 98%

Supplier: Ambeed
Danger

Synonyms: Platinum (II) bis(2,4-pentanedionato-O,O'), Platinum (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')platinum (II)

A394836-100MG A394836-1G A394836-250MG A394836-5G A394836-25G
76725-020EA 13.8 USD
76725-020 76725-022 76725-024 76725-026 77551-180
Platinum(II) acetylacetonate 98%
Platinum(II) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄Pt
MW: 393.3 g/mol
MDL Number: MFCD00000028
CAS Number: 15170-57-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR