Acenaphthenequinone 98%

Supplier: Ambeed
Warning

A394664-100G A394664-1G A394664-25G A394664-500G A394664-5G
76725-010EA 202 USD
76725-010 76725-012 76725-014 76725-016 76725-018
Acenaphthenequinone 98%
Acenaphthenequinone

Precaución: for Research Use Only

Formula: C₁₂H₆O₂
MDL Number: MFCD00003805
CAS Number: 82-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR