3,3'-Difluorobenzophenone 97%

Supplier: Ambeed
Warning

A394209-1G A394209-250MG A394209-25G A394209-5G
76725-002EA 21 USD
76725-002 76725-004 76725-006 76725-008
3,3'-Difluorobenzophenone 97%
3,3'-Difluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O
MDL Number: MFCD00000337
CAS Number: 345-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR