3-Nitro-1,8-naphthalic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitrobenzo[de]isochromene-1,3-dione

A393514-100G A393514-1G A393514-25G A393514-5G
76724-994EA 526.21 USD
76724-994 76724-996 76724-998 76725-000
3-Nitro-1,8-naphthalic anhydride 98%
3-Nitro-1,8-naphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₅NO₅
MW: 243.1718 g/mol
MDL Number: MFCD00006926
CAS Number: 3027-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR