Palmitic anhydride 96%

Supplier: Ambeed
Danger

A392769-5G A392769-25G
76724-986EA 15 USD
76724-986 77879-847
Palmitic anhydride 96%
Palmitic anhydride

Precaución: for Research Use Only

Formula: C₃₂H₆₂O₃
MDL Number: MFCD00008992
CAS Number: 623-65-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR