Octyl-β-D-thioglucopyranoside 99%

Supplier: Ambeed
Warning

Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

A391799-1G A391799-25G A391799-5G
76724-978EA 14.86 USD
76724-978 76724-980 76724-982
Octyl-β-D-thioglucopyranoside 99%
Octyl β-D-thioglucopyranoside

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR