N-(1-Phenyl-ethyl)-acetamide 98%

Supplier: Ambeed
Warning

Synonyms: N-(1-Phenylethyl)acetamide

A391002-10G A391002-1G A391002-5G
76724-974EA 230.22 USD
76724-974 76724-976 77668-802
N-(1-Phenyl-ethyl)-acetamide 98%
N-(1-Phenyl-ethyl)-acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.216 g/mol
MDL Number: MFCD00040690
CAS Number: 36065-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR