2-Fluoro-5-nitrophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-nitrophenylboronic acid, pinacol ester, 2-(2-Fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-nitrobenzeneboronic acid pinacol ester

A390914-10G A390914-1G A390914-250MG A390914-5G A390914-25G
76724-966EA 53 USD
76724-966 76724-968 76724-970 76724-972 77879-843
2-Fluoro-5-nitrophenylboronic acid pinacol ester 98%
2-Fluoro-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BFNO₄
MDL Number: MFCD06409955
CAS Number: 425378-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR