cis-N-Boc-4-cyano-L-proline methyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-cis-4-CN-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, 1-tert-Butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate
A390780-1G
A390780-5G
A390780-250MG
76724-964EA
11
USD
76724-964
77594-194
77601-718
cis-N-Boc-4-cyano-L-proline methyl ester 98%
cis-N-Boc-4-cyano-L-proline methyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂O₄ |
MDL Number:
MFCD03787928 CAS Number: 487048-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...