N-(4-Iodophenyl)cyclopropanecarboxamide 95%

Supplier: Ambeed
Warning

A388186-10G A388186-1G A388186-250MG A388186-5G
76724-944EA 177.18 USD
76724-944 76724-946 76724-948 76724-950
N-(4-Iodophenyl)cyclopropanecarboxamide 95%
N-(4-Iodophenyl)cyclopropanecarboxamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀INO
MDL Number: MFCD01327898
CAS Number: 23779-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR